Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Respiratory Protection
▸
Air-Purifying Respirators
▸
Powered Air Purifying Respirators
▸
PAPR Replacement Parts and Accessories
▸
PAF-1024 Car Charger
Actual product may vary
CleanSpace
PAF-1024 Car Charger
Car Charger designed for use with all CleanSpace2™ PAPR models.
Have Questions?
Catalog Number
For Use With (Equipment)
CSEPAF-1024
CleanSpace2™
Catalog number
CSEPAF-1024
Price (SGD)
84.00
Each
Add to cart
For Use With (Equipment):
CleanSpace2™
Price (SGD)
84.00
Each
Add to cart
Black
For use with all CleanSpace2™ Respirator
Specifications
Color
Black
Product Type
Car Charger
For Use With (Equipment)
CleanSpace2™
Model
PAF-1024