Full Face Mask Spectacle Kit
Full Face Mask Spectacle Kit
Actual product may vary
CleanSpace Respirators
Full Face Mask Spectacle Kit
Full face mask spectacle kit is designed to allow prescription lens' to be used without compromising the face seal.
Have Questions?
Catalog number CSEPAF-1017
Price (SGD)
391.00
EA
Add to cart
Price (SGD)
391.00
EA
Add to cart
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
  • The frame is fully adjustable for all face sizes, wide frames to fit large lenses for peripheral vision
  • The frame is engineered to provide stability during vigorous activity and is easy to fit and adjust
Specifications
Certifications/ComplianceAS/NZS1716: 2012, EN 12942
DisposableReusable
MaterialAluminum, Silicone
Product TypeSpectacle Kit
For Use With (Application)Welding, Woodworking, Manufacturing, Smelting, Construction, Recycling Plants, Emergency Services, Mining, Agriculture, Processing Plants, Grinding
For Use With (Equipment)PAF-1014, PAF-1106
ModelPAF-1017
Storage RequirementsStore at -10°C to +55°C
Dimensions (L x W x H)70 x 150 x 70 mm
Weight (Metric)150 g
Unit SizeEA