Ultra Decontamination Plug Set
Actual product may vary
CleanSpace Respirators
Ultra Decontamination Plug Set
For use with the CleanSpace™ Ultra Respirator unit (PAF-0070). Seals all the openings on the CleanSpace™ Ultra PAPR unit for storage during and after decontamination. The Plug Set is made of nylon and silicone and is safe for use in decontamination operations.
Have Questions?
Catalog NumberFor Use With (Equipment)
CSEPAF-0074CleanSpace™ Ultra Respirator unit (PAF-0070)
Catalog number CSEPAF-0074
Price (SGD)
134.00
Each
For Use With (Equipment):
CleanSpace™ Ultra Respirator unit (PAF-0070)
Price (SGD)
134.00
Each
  • Used with the revolutionary CleanSpace Ultra PAPR: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
Specifications
MaterialNylon, Silicone
For Use With (Equipment)CleanSpace™ Ultra Respirator unit (PAF-0070)
Dimensions (L x W x H)800 x 180 x 110 mm
Weight (Metric)100 g
Unit SizeEach