Intrinsically safe battery-powered, positive pressure air-purifying respirator (PAPR). Suitable for use in certain ignitable or potentially explosive atmospheres where there is a risk of explosion.
Have Questions?
Catalog number CSEPAF-0060
Price (SGD)
2,523.00
Each
Price (SGD)
2,523.00
Each
Intrinsically safe
Light weight and comfortable
High visibility yellow casing for easy recognition
AirSensit System™ for mask pressure control and breath responsive airflow
One-button system and standby and auto-start/stop mode for quick don/doffing
Easy to use support harness
Altitude compensated filter blocked detection
Long operating time: up to 8 hours and quick battery recharging: < 2hours