Large Case Particulate Pre-Filters
Actual product may vary
Large Case Particulate Pre-Filters
Suitable for protection against airborne particulates (dust, mists and fumes). Designed to remove course particles and enhance the life of the Particulate P3 Filter on PAF-0037 and Combined Filters. The Pre-Filter should be changed after each use.
Have Questions?
Catalog number CSEPAF-0057
Price (SGD)
73.00
Pack
Price (SGD)
73.00
Pack
  • Used with the revolutionary CleanSpace - a light weight PAPR with no hoses/belts
  • Materials: Spun polymer fibers
  • Easy and quickly fitted and removed from the power unit
  • Sold in packs of 20
  • Compatible with all CleanSpace Respirators
Specifications
Product TypePrefilter
Unit SizePack