Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Respiratory Protection
▸
Air-Purifying Respirators
▸
Powered Air Purifying Respirators
▸
PAPR Cartridges and Filters
▸
Combined Filter
Actual product may vary
CleanSpace
Combined Filter
Combined filter suitable for protection against airborne particulate (dust, mists, and fumes) and organic gases or vapors (boiling point > 65°C).
Have Questions?
Catalog Number
Model
CSEPAF-0050
A1 P3 TM3 SLR
Catalog number
CSEPAF-0050
Price (SGD)
109.00
Each
Add to cart
Model:
A1 P3 TM3 SLR
Price (SGD)
109.00
Each
Add to cart
Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
Materials: Fibreglass particulate media, activated carbon and plastic casing, silicone seal
Easily fitted and removed from the power unit
Specifications
Certifications/Compliance
EN 12942
Product Type
Filter
For Use With (Equipment)
CleanSpace PAPR Power Units
Model
A1 P3 TM3 SLR