PAPR Half Mask
Actual product may vary
CleanSpace Respirators
PAPR Half Mask
Half masks clip quickly onto PAPR power units and easily unclip for cleaning. Specifically designed to be used with the CleanSpace™ 2, Ultra and EX PAPR systems.
Have Questions?
Catalog NumberSize
CSEPAF-0033Small
Catalog number CSEPAF-0033
Price (SGD)
229.00
Each
Size:
Small
Price (SGD)
229.00
Each
  • Compatible with ALL CleanSpace™PAPR power units
  • Used with the revolutionary CleanSpace™PAPR: lightweight, no hoses or belts
  • Available in 3 sizes to facilitate a secure seal and a comfortable fit
  • Materials: Soft silicone cushion, nylon clips and polycarbonate (exhalation valve)
  • Easy removal for cleaning
  • Transparent and facilitates communication
  • Clip quickly onto the power units and are easily unclipped for cleaning
  • Soft silicone that readily moulds to the face and does not cause allergic dermatitis
Specifications
Certifications/ComplianceAS/NZS1716: 2012
Certifications (EN)EN 12942
Product TypePAPR Half Mask
SizeSmall
Facepiece/Headtop ModelPAPR half mask only
For Use With (Application)Mining, welding, manufacturing, smelting, construction, recycling plants, emergency services, agriculture, processing plants, grinding
For Use With (Equipment)PAF-1034, PAF-2034
IP RatingIP66/67
ModelPAF-0033
Storage RequirementsStore at -10°C to 55°C
Dimensions (L x W x H)170 x 130 x 70 mm
Weight (Metric)125 g
Unit SizeEach