HydroPure™ with Purified Water
Actual product may vary
Contec
HydroPure™ with Purified Water
HydroPure™ with Purified Water designed for cleaning of life science cleanrooms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberContainer TypeQuantity
CNTSBC56HPCapped Bottle2 x 5 L
CNTSBT16HPWTrigger Spray Bottle6 x 1 L
Catalog number CNTSBC56HP
Price (SGD)
399.00
Case
Container Type:
Capped Bottle
Quantity:
2 x 5 L
Price (SGD)
399.00
Case
Designed for cleaning of life science cleanrooms.
  • Composition: 6% hydrogen peroxide in water
  • Can be applied directly to surface ensuring even coverage
  • Filtered to 0.22 microns under Grade A airflow
  • Physically removes contaminants
  • Sufficient sporicidal activity for Grade A / B area
  • Low residue, eliminates need for residue removal stage with alcohol or water
  • Gamma irradiated
  • Large droplet size
  • Facilitates transfer disinfection into isolators and RABS
  • Same grade as when vaporized hydrogen peroxide is used
  • Triple / double bagged in linear tear packaging
  • Noncorrosive
Applications:
  • Efficacious against bacteria, fungi, moulds, yeasts and spores
Specifications
Product TypeDetergent with Purified Water
FormLiquid
pH6.5 to 7.5
SterilitySterile
AppearanceColorless
AutoclavableNot Autoclavable
ColorClear
CompositionHydrogen peroxide and purified water (EP)
Concentration6% Hydrogen peroxide in purified water
Container TypeCapped Bottle
DisposableLimited-use
FilteredFiltered
FlammableNo
For Use With (Application)Used in pharmaceutical industry
Quantity2 x 5 L
Shelf Life1 Year
Volume (English)136 oz.
Volume (Metric)5 L
Unit SizeCase