HydroPure
HydroPure
Actual product may vary
HydroPure
Blend of 6% hydrogen peroxide and purified water, low residue, sporicidal biocide. HydroPure is filtered to 0.2μ leaves no residue, eliminating any residue removal stage.
Have Questions?
Catalog NumberQuantityContainer Type
CNTFBT16HP6 x 1 LTrigger Spray Bottle
Catalog number CNTFBT16HP
Price (SGD)
227.00
EA
Add to cart
Quantity:
6 x 1 L
Container Type:
Trigger Spray Bottle
Price (SGD)
227.00
EA
Add to cart
Blend of 6% hydrogen peroxide and purified water, low residue, sporicidal biocide. Contec™ HydroPure is filtered to 0.2μ leaves no residue, eliminating any residue removal stage.
  • Efficacious against bacteria, fungi, moulds, yeasts and spores
  • 0.2μm filtered and filled under Grade A uni-directional flow air and bagged in Grade B cleanroom
  • Leaves little to no residue and has endotoxin level of <0.25 EU/mL
  • Safe to use in product contact areas
  • Designed for ease of entry into pharmaceutical cleanrooms
  • Ensures the product is free from contamination and particulates
  • Sufficient sporicidal activity for Grade A/B area
  • Large droplet size reduces the risk of inhalation and provides good surface coverage
  • Bagged in polyethylene linear tear bags
  • Facilitates transfer disinfection into isolators and RABS
  • RTU with one year shelf life
Specifications
Product TypeHydrogen Peroxide and Purified Water Blend
FormLiquid
pHNot Applicable
ScentUnscented
SterilityNon-sterile
AppearanceClear
AutoclavableNot Autoclavable
ColorClear
Composition6% hydrogen peroxide in purified water
Container TypeTrigger Spray Bottle
DisposableLimited-use
FilteredFiltered
FlammableYes
Quantity6 x 1 L
Shelf Life1 Year
Volume (Metric)1 L
Unit SizeEA