Owl™ Gel-Drying Supplies
Owl™ Gel-Drying Supplies
Actual product may vary
Owl™ Gel-Drying Supplies
Use the frames and pre-cut cellophane that come in Thermo Scientific™ Owl™ Gel-Drying Supplies with Owl™ Gel-Drying Kits.
Have Questions?
Catalog NumberProduct TypeLength (Metric)Width (Metric)
CM-10Cellophane, Precut14 cm14 cm
Catalog number CM-10
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
Cellophane, Precut
Length (Metric):
14 cm
Width (Metric):
14 cm
Combine the frames and pre-cut cellophane that come in Thermo Scientific™ Owl™ Gel-Drying Supplies with Owl™ Gel-Drying Kits.
  • Frames and pre-cut cellophane available in large (24 × 24cm) or small (14 × 14cm) format
Specifications
Gel Size14 x 14 cm
Product TypeCellophane, Precut
Length (Metric)14 cm
Width (Metric)14 cm
Unit SizeEA