Hastelloy External Probe
Hastelloy External Probe
Actual product may vary
Hastelloy External Probe
For Use With: Isotemp Hotplates and Hotplate Stirrers
Have Questions?
Catalog NumberFor Use With (Equipment)
CIC0001467Isotemp Hotplates and Hotplate Stirrers
Catalog number CIC0001467
Price (SGD)
321.00
EA
Add to cart
For Use With (Equipment):
Isotemp Hotplates and Hotplate Stirrers
Price (SGD)
321.00
EA
Add to cart
Hastelloy PT1000 Temperature probe for use with Isotemp and Isotemp Advanced hotplates and hotplate stirrers
  • Hastelloy PT1000 Temperature probe
  • Temp. Range: 0 to 400°C
  • Chemical and corrosion resistant
Specifications
For Use With (Equipment)Isotemp Hotplates and Hotplate Stirrers
Temperature (Metric)0°C to 400°C
TypeProbe
Unit SizeEA