CEN5p (R6G) FISH Probe
CEN5p (R6G) FISH Probe
Actual product may vary
CEN5p (R6G) FISH Probe
Labeled FISH probes as chromosomal markers using Fluorescent In Situ Hybridization Technique.
Have Questions?
Catalog NumberQuantity
BSIFC-003820 μL
Catalog number BSIFC-0038
Price (SGD)
-
Quantity:
20 μL
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Label or DyeR6G
For Use With (Application)Fluorescent In Situ Hybridization (Cell)
IncludesFISH Hybridization Buffer (80 uL)
Location5p12
ProbeCEN5p
Product TypeCEN5p (R6G) FISH Probe
Quantity20 μL
Unit SizeEA
Contents & Storage
Store at 4°C in the dark.