Knitted Polyester Lab-Tips Swabs
Knitted Polyester Lab-Tips Swabs
Actual product may vary
Knitted Polyester Lab-Tips Swabs
For cleaning grooved, slotted and recessed areas
Have Questions?
Catalog NumberTip StyleLength (Metric) HandleLength (Metric) Tip
BERLTP70R.20Rigid58 mm12.5 mm
Catalog number BERLTP70R.20
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Tip Style:
Rigid
Length (Metric) Handle:
58 mm
Length (Metric) Tip:
12.5 mm
  • Continuous filament polyester knit head
  • Short polypropylene handle with a rigid head
  • Thermally bonded head
  • Laundered and packaged in Class 10 (ISO 4) cleanroom
  • Autoclavable
Specifications
Certifications/ComplianceISO Class 4
Diameter (English) Handle0.11 in.
Length (English)2.8 in.
For Use With (Application)Designed for the highest level of contamination control in critical processing applications, cleaning of sensitive equipment, applying lubricants and other liquids, removing excess materials
Handle ColorGreen
Product TypeSwab
Diameter (Metric) Handle2.8 mm
Length (Metric)70 mm
Length (English) Handle2.28 in.
Length (Metric) Handle58 mm
Thickness (Metric)3.8 mm
Length (English) Tip0.49 in.
Length (Metric) Tip12.5 mm
Width (Metric) Tip3.8 mm
Width (English) Tip0.15 in.
Tip StyleRigid
Tip MaterialPolyester
Unit SizeCS