Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems
Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems
Actual product may vary
Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems
Cast and run a gel in the buffer chamber and achieve flat, even banding patterns on a consistent basis with these B1A Mini Gel Electrophoresis systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
B1A-BPEasyCast w/ Buffer Exchange Ports
B1AEasyCast
Catalog number B1A-BP
Price (SGD)
1,052.00
EA
Add to cart
Includes:
EasyCast w/ Buffer Exchange Ports
Price (SGD)
1,052.00
EA
Add to cart
Cast and run a gel in the buffer chamber and achieve flat, even banding patterns on a consistent basis with Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems. The ideal size for every lab, this system determines molecular weight ranges of PCR fragments or larger sized DNA molecules.
Features:
  • Two comb slots on gel tray double sample capacity by doubling number of sample wells.
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.
  • Leakproof casting allows a gel to be cast and run in the same chamber with no tape or additional parts required
  • Run from five to 24 samples in 15 minutes.
Ordering Information:
Available with optional buffer exchange ports (B1A-BP) for use with an external recirculation pump.
Includes:
  • Main buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Thermo Scientific™ EasyCast gasketed UVT gel tray
  • Two heavy-duty, one-piece, double-sided combs: 6 & 10 well, 1.0/1.5mm thick
Specifications
Certifications/ComplianceCE marked
Gel SizeMini
Product LineOwl
TypeMini Gel Electrophoresis System
IncludesEasyCast w/ Buffer Exchange Ports
Sample Size5 to 24
Volume (Metric) Buffer400 mL
Dimensions (L x W x H)6.31 x 4.13 x 3.75 in. (16 x 10.5 x 9.5 cm)
Length (English)6.31 in.
Length (Metric)16 cm
Length (Metric) Gel8 cm
Width (English)4.13 in.
Width (Metric)10.5 cm
Width (Metric) Gel7 cm
Height (English)3.75 in.
Height (Metric)9.5 cm
Unit SizeEA
Frequently asked questions (FAQs)
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.