Sterile Plastic Inoculation Loops (Volume: 5μL)
Sterile Plastic Inoculation Loops (Volume: 5μL)
Actual product may vary
Sterile Plastic Inoculation Loops (Volume: 5μL)
Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture
Have Questions?
Catalog NumberQuantityPackaging TypeSterility
B150 Pouches of 20Grip Seal BagAseptically Manufactured
Catalog number B1
Price (SGD)
-
Quantity:
50 Pouches of 20
Packaging Type:
Grip Seal Bag
Sterility:
Aseptically Manufactured
  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Medium rigidity
  • Culture loops (Length: 200mm)
  • Smooth edges to avoid tearing and digging in agar
  • Volume: 5μL
  • Color: Blue
Specifications
DisposableSingle-use
Diameter (English) Inside0.10 in.
Diameter (Metric) Inside2.5 mm
Length (English)7.87 in.
Length (Metric)200 mm
MaterialHigh-impact Polystyrene
SterilityAseptically Manufactured
ColorBlue
Product TypePlastic Culture Loops
Length (English) Handle7.87 in.
Length (Metric) Handle200 mm
Quantity50 Pouches of 20
Packaging TypeGrip Seal Bag
CalibratedYes
Unit SizeEA
Frequently asked questions (FAQs)
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.