Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Cold Storage Products
▸
Refrigerator and Freezer Accessories
▸
CryoClaw
Actual product may vary
Thermo Scientific
CryoClaw
Retrieve vials, ampoules and cryoboxes from cryogenic vessels using the Thermo Scientific™ CryoClaw.
Have Questions?
Catalog Number
For Use With (Equipment)
AY509X18
All CryoPlus Models
Catalog number
AY509X18
HSN
82032000
Price (SGD)
383.00
Each
Add to cart
For Use With (Equipment):
All CryoPlus Models
Price (SGD)
383.00
Each
Add to cart
Retrieve vials, ampoules and cryoboxes from cryogenic vessels using the Thermo Scientific™ CryoClaw.
Rubber suction cups retrieve ampules or boxes that fall from racks or cans into cryogenic vessels
Can also be used to reach overhead items or to safely move radioisotopes without touching containers
Length 41in. (104cm)
Specifications
Description
CryoClaw Ampule Remover
Type
CryoClaw
For Use With (Equipment)
All CryoPlus Models
Holds
Vials