Eurotubo Polystyrene Centrifuge Tubes with Cap
Actual product may vary
As One
Eurotubo Polystyrene Centrifuge Tubes with Cap
These free-standing Eurotubo Polystyrene Centrifuge Tubes without Cap, with external skirt, can be autoclaved with loose fitting cap. Supplied with pre-fitted cap.
Have Questions?
Catalog NumberCapacity (Metric)
ASO3014-0312 mL
Catalog number ASO3014-03
Price (SGD)
-
Capacity (Metric):
12 mL
  • Samples and/or urine collection containers
  • Autoclavable
  • External skirt
  • Green cap
Specifications
ColorClear
Capacity (Metric)12 mL
MaterialPolystyrene
No. per Case1560/Cs.
No. per Pack250 Pk.
PackagingCase
AutoclavableAutoclavable
Length (Metric)103 mm
Bottom ShapeRound
Diameter (Metric)16.3 mm
Product TypeCentrifuge Tube
Max. RCF7500 x g
Unit SizePieces