TouchNTuff™ 92-605 Disposable Chemical Resistant Nitrile Gloves
Actual product may vary
TouchNTuff™ 92-605 Disposable Chemical Resistant Nitrile Gloves
Disposable, nitrile gloves with extended cuff features splash protection against hazardous chemicals.
Have Questions?
Catalog NumberSize
ANL92-605-L-BX7.5/8
Catalog number ANL92-605-L-BX
Price (SGD)
241.00
Box
Size:
7.5/8
Price (SGD)
241.00
Box
Disposable, nitrile gloves with extended cuff features splash protection against hazardous chemicals.
  • Does not contain natural rubber latex proteins
  • Strong grip in wet or dry conditions
  • Rolled beaded for easy donning
  • Powder free, Polymer-coated
  • Ambidextrous
  • Textured Fingers
  • AQL: 1.5
Specifications
ChlorinatedYes
AQL1.5
ColorGreen
DisposableSingle-use
Finger Thickness (English)6.2 mil
Finger Thickness (Metric)0.16 mm
Latex FreeYes
Palm Thickness (English)4.7 mil
Palm Thickness (Metric)0.12 mm
Size7.5/8
EN 421Yes
EN ISO 374-1Type B
Glove Cuff StyleBeaded/Rolled
Unit SizeBox