AlphaTec™ 15 in. Solvex™ 37-165 Nitrile Gloves
Actual product may vary
AlphaTec™ 15 in. Solvex™ 37-165 Nitrile Gloves
AlphaTec™ 15 in. Solvex™ 37-165 Nitrile Gloves
AlphaTec™ 15 in. Solvex™ 37-165 Nitrile Gloves
AlphaTec™ 15 in. Solvex™ 37-165 Nitrile Gloves
AlphaTec™ 15 in. Solvex™ 37-165 Nitrile Gloves
Heavy-duty 22 mil nitrile gloves offer versatile chemical protection that performs across a diverse range of applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
ANL37-165-909
ANL37-165-808
Catalog number ANL37-165-90
Price (SGD)
0.00
Pair
Size:
9
Price (SGD)
0.00
Pair
Heavy-duty 22 mil nitrile gloves offer versatile chemical protection that performs across a diverse range of applications.
  • Nitrile construction protects against abrasions, cuts, snags and punctures
  • Curved finger design and contoured palm help reduce fatigue
  • Full cut for easy donning and doffing
  • Does not contain protein allergens
  • No inner cotton flocking, reducing risk of lint contamination
  • Sandpatch finish gives the glove a smooth surface
  • Fully reusable, with an unequaled abrasion resistance
  • Complies with FDA food handling requirements (21 CFR 177.2600)
Specifications
ANSI Abrasion RatingANSI 5
Certifications/ComplianceFDA 21 CFR 177.2600
Certifications/Compliance (Europe)CE 0493, EN 420:2003 + A1:2009, Category III
ChlorinatedYes
ColorGreen
Latex FreeYes
MaterialNitrile
Size9
Thickness (English)22 mil
Thickness (Metric)0.55 mm
EN388 StandardEN 388:4102, EN 388:4102X
EN 421No
EN ISO 374-1Type A
EN ISO 374-1 CHEMICAL LETTER CODESAGJKLPT
EN ISO 374 - 5Bacteria, Fungi only
For Use With (Application)Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Glove Cuff StyleGauntlet
Grip DesignSandpatch
LiningUnlined
Model37-165 9 117209
SeriesAlphaTec™ Solvex™ 37-165
Length (English)15 in.
Length (Metric)38 cm
Unit SizePair