Ethanol, 99.8%, for HPLC, absolute
Ethanol, 99.8%, for HPLC, absolute
Ethanol, 99.8%, for HPLC, absolute
Thermo Scientific Chemicals
Ethanol, 99.8%, for HPLC, absolute
CAS: 64-17-5 | C2H6O | 46.07 g/mol
Have Questions?
Catalog NumberQuantity
ACO4457400252.5 L
Catalog number ACO445740025
Price (SGD)
638.00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (SGD)
638.00
Each
Specifications
Chemical Name or MaterialEthanol, absolute
CAS64-17-5
Health Hazard 1Danger
CAS7732-18-5
Quantity2.5 L
View more
For Manufacturing and Laboratory Use. Not for beverage use