Ethanol, 70%, technical, not denaturated
Thermo Scientific Chemicals
Ethanol, 70%, technical, not denaturated
CAS: 64-17-5 | C2H6O | 46.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO4442800505 LPlastic Drum
Catalog number ACO444280050
Price (SGD)
668.00
Each
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (SGD)
668.00
Each
Specifications
Chemical Name or MaterialEthanol
Melting Point-48.0°C
CAS64-17-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Highly flammable liquid and vapor.
Causes serious eye irritation.
View more
For Manufacturing and Laboratory Use. Not for beverage use