Heptanes, technical, mixture of isomers
Heptanes, technical, mixture of isomers
Heptanes, technical, mixture of isomers
Heptanes, technical, mixture of isomers
Thermo Scientific Chemicals
Heptanes, technical, mixture of isomers
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO4442700505 LPlastic Drum
Catalog number ACO444270050
Price (SGD)
94.40
Special offer
Ends: 31-Mar-2026
118.00
Save 23.60 (20%)
EA
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (SGD)
94.40
Special offer
Ends: 31-Mar-2026
118.00
Save 23.60 (20%)
EA
Add to cart
Specifications
Chemical Name or MaterialHeptanes
CAS142-82-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapour.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap