Isopropanol, technical
Isopropanol, technical
Isopropanol, technical
Thermo Scientific Chemicals
Isopropanol, technical
CAS: 67-63-0 | C3H8O | 60.1 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO4442500505 LPlastic drum
Catalog number ACO444250050
Price (SGD)
192.00
Each
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (SGD)
192.00
Each
Specifications
Chemical Name or MaterialIsopropanol
CAS67-63-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present