hydrogen peroxide, 3 wt.% solution in water, stabilized
hydrogen peroxide, 3 wt.% solution in water, stabilized
hydrogen peroxide, 3 wt.% solution in water, stabilized
hydrogen peroxide, 3 wt.% solution in water, stabilized
Thermo Scientific Chemicals
hydrogen peroxide, 3 wt.% solution in water, stabilized
CAS: 7722-84-1 | H2O2 | 34.014 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Plastic bottle
Catalog number ACO426001000
Price (SGD)
41.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
41.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydrogen peroxide
Name Note3 wt.% solution in water, stabilized
CAS7722-84-1
Boiling Point100.0°C
Health Hazard 2GHS H Statement:
Causes skin irritation.
View more
Oxidation reagent
RUO – Research Use Only