1x1 image pixel for data collection
Zinc sulfate heptahydrate, 99%, ACS reagent
Zinc sulfate heptahydrate, 99%, ACS reagent
Zinc sulfate heptahydrate, 99%, ACS reagent
Zinc sulfate heptahydrate, 99%, ACS reagent
Thermo Scientific Chemicals
Zinc sulfate heptahydrate, 99%, ACS reagent
CAS: 7446-20-0 | H14O11SZn | 287.54 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO424605000500 gPlastic bottle
Catalog number ACO424605000
Price (SGD)
93.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
93.00
EA
Add to cart
Specifications
Chemical Name or MaterialZinc sulfate heptahydrate
Melting Point100.0°C
CAS7446-20-0
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
RUO – Research Use Only
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link