Tin(II) chloride dihydrate, 98+%, ACS reagent
Tin(II) chloride dihydrate, 98+%, ACS reagent
Tin(II) chloride dihydrate, 98+%, ACS reagent
Tin(II) chloride dihydrate, 98+%, ACS reagent
Thermo Scientific Chemicals
Tin(II) chloride dihydrate, 98+%, ACS reagent
CAS: 10025-69-1 | Cl2Sn·2H2O | 225.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic Bottle
Catalog number ACO424481000
Price (SGD)
142.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (SGD)
142.00
EA
Add to cart
Specifications
Boiling Point652.0°C
Chemical Name or MaterialTin(II) chloride dihydrate
Melting Point37.0°C to 38.0°C
CAS10025-69-1