Sodium bicarbonate, 99.7+%, ACS reagent
Sodium bicarbonate, 99.7+%, ACS reagent
Sodium bicarbonate, 99.7+%, ACS reagent
Sodium bicarbonate, 99.7+%, ACS reagent
Thermo Scientific Chemicals
Sodium bicarbonate, 99.7+%, ACS reagent
CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO4242700101 kgPlastic bottle
Catalog number ACO424270010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium bicarbonate
Melting Point270.0°C
CAS144-55-8
ColorWhite
Physical FormPowder