View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBoric acid
ColorBrown-Red
CAS10043-35-3
Health Hazard 1Danger
Health Hazard 2GHS H Statement: May damage fertility. May damage the unborn child.