Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
Thermo Scientific Chemicals
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
CAS: 26837-42-3 | (C2H3KO4S)n | NaN g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO41814025025 gGlass bottle
Catalog number ACO418140250
Price (SGD)
1,499.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
1,499.00
EA
Add to cart
Specifications
Chemical Name or MaterialPolyvinylsulfuric acid potassium salt
ColorBeige-Brown to White
CAS26837-42-3
Physical FormCrystalline Powder
Molecular Formula(C2H3KO4S)n