1x1 image pixel for data collection
Alizarin Red S, pure, certified
Alizarin Red S, pure, certified
Alizarin Red S, pure, certified
Alizarin Red S, pure, certified
Thermo Scientific Chemicals
Alizarin Red S, pure, certified
CAS: 130-22-3 | C14H7NaO7S | 342.253 g/mol
Have Questions?
Catalog NumberQuantity
ACO400481000100 g
Catalog number ACO400481000
Price (SGD)
372.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
372.00
EA
Add to cart
Specifications
Chemical Name or MaterialAlizarin Red S
CAS130-22-3
Health Hazard 1Warning
Absorption Ratio0.97 to 1.07 (for the first maximum) (P-15/P+15)
RTECS NumberCB1095300
View more
Indicator
RUO – Research Use Only
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link