Ammonium hydroxide, for analysis, 25% solution in water
Ammonium hydroxide, for analysis, 25% solution in water
Ammonium hydroxide, for analysis, 25% solution in water
Ammonium hydroxide, for analysis, 25% solution in water
Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 25% solution in water
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO3900300252.5 LGlass bottle
Catalog number ACO390030025
Price (SGD)
153.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
153.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium hydroxide
ColorColorless
Boiling Point38.0°C
CAS1336-21-6
Health Hazard 1Danger