1x1 image pixel for data collection
Titanium(IV) oxide, Aeroxide™ P25
Titanium(IV) oxide, Aeroxide™ P25
Titanium(IV) oxide, Aeroxide™ P25
Titanium(IV) oxide, Aeroxide™ P25
Titanium(IV) oxide, Aeroxide™ P25
Thermo Scientific Chemicals
Titanium(IV) oxide, Aeroxide™ P25
CAS: 13463-67-7 | O2Ti | 79.87 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO384292500250 gPlastic bottle
Catalog number ACO384292500
Price (SGD)
215.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
215.00
EA
Add to cart
Specifications
Boiling Point2900.0°C
Chemical Name or MaterialTitanium(IV) oxide
Melting Point1855.0°C
CAS13463-67-7
ColorWhite to Yellow
View more
Suitable for battery materials development.
RUO – Research Use Only
Share catalog number, name or link