Stearyl stearate, 98%
Stearyl stearate, 98%
Stearyl stearate, 98%
Stearyl stearate, 98%
Thermo Scientific Chemicals
Stearyl stearate, 98%
CAS: 2778-96-3 | C36H72O2 | 536.97 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO3681100505 gGlass bottle
Catalog number ACO368110050
Price (SGD)
1,180.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
1,180.00
EA
Add to cart
Specifications
Chemical Name or MaterialStearyl stearate
Name Note98%
CAS2778-96-3
ColorWhite
EINECS Number220-476-5