1x1 image pixel for data collection
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™
N,N-Dimethylformamide, 99.8%, C3H7NO, CAS Number-68-12-2, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dwumetyloformamid, dimethylformamide, dmfa, dimethyl formamide, dimetilformamide, dmf, dimethylformamid, 1L, 0.01 % max., Vapor Pressure: 4.9mbar at 20 deg.C, 04, 58 | CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACO348431000
Price (SGD)
110.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (SGD)
110.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N-Dimethylformamide
Melting Point-61.0°C
Name Notewater <50 ppm
CAS68-12-2
CAS Min %99.75
View more
RUO – Research Use Only
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link