Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACO327182500250 mLGlass Bottle
Catalog number ACO327182500
Price (SGD)
212.00
Special offer
Ends: 31-Mar-2026
265.00
Save 53.00 (20%)
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
212.00
Special offer
Ends: 31-Mar-2026
265.00
Save 53.00 (20%)
EA
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide
Melting Point18.4°C
CAS67-68-5
Health Hazard 1HYGROSCOPIC
Density1.1000g/mL