Methanol, 99.9%, for HPLC gradient grade
Methanol, 99.9%, for HPLC gradient grade
Methanol, 99.9%, for HPLC gradient grade
Thermo Scientific Chemicals
Methanol, 99.9%, for HPLC gradient grade
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO3257400252.5 LGlass Bottle
Catalog number ACO325740025
Price (SGD)
124.00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
124.00
Each
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Filtered to 0.2 micrometerFiltered
For HPLC
RUO – Research Use Only