Potassium ferricyanide, 99+%, for analysis
Potassium ferricyanide, 99+%, for analysis
Potassium ferricyanide, 99+%, for analysis
Potassium ferricyanide, 99+%, for analysis
Thermo Scientific Chemicals
Potassium ferricyanide, 99+%, for analysis
CAS: 13746-66-2 | C6FeK3N6 | 329.25 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO2231100252.5 kgPlastic bottle
Catalog number ACO223110025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium ferricyanide
Name Notep.a.
CAS13746-66-2
ColorOrange to Red
Health Hazard 1Warning
View more
Indicator
RUO – Research Use Only