Acetic acid, sodium salt, 99+%, for analysis, anhydrous
Acetic acid, sodium salt, 99+%, for analysis, anhydrous
Acetic acid, sodium salt, 99+%, for analysis, anhydrous
Acetic acid, sodium salt, 99+%, for analysis, anhydrous
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO220892500250 gPlastic bottle
Catalog number ACO220892500
Price (SGD)
70.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
70.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetic acid, sodium salt
Name Notep.a.
pH7.5 to 9.2 (5% soln. at 20°C)
CAS127-09-3
Melting Point324.0°C
View more
Biological buffer
RUO – Research Use Only