1x1 image pixel for data collection
Ammonium sulfate, 99.9995%, (trace metal basis)
Ammonium sulfate, 99.9995%, (trace metal basis)
Ammonium sulfate, 99.9995%, (trace metal basis)
Ammonium sulfate, 99.9995%, (trace metal basis)
Thermo Scientific Chemicals
Ammonium sulfate, 99.9995%, (trace metal basis)
CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO194152500250 gGlass bottle
Catalog number ACO194152500
Price (SGD)
724.00
EA
Estimated availability date 24-Sep-2025
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
724.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium sulfate
Melting Point280.0°C
CAS7783-20-2
ColorYellow to Orange
Health Hazard 1GHS Signal Word: Warning
View more
For ultra trace analysis
RUO – Research Use Only
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link