(+)-Limonene, 96%, unstabilized
(+)-Limonene, 96%, unstabilized
(+)-Limonene, 96%, unstabilized
(+)-Limonene, 96%, unstabilized
Thermo Scientific Chemicals
(+)-Limonene, 96%, unstabilized
CAS: 5989-27-5 | C10H16 | 136.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1793900252.5 LGlass bottle
Catalog number ACO179390025
Price (SGD)
425.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
425.00
EA
Add to cart
Specifications
Chemical Name or Material(+)-Limonene
CAS5989-27-5
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0