Glycerol, 99+%, Extra Pure
Glycerol, 99+%, Extra Pure
Glycerol, 99+%, Extra Pure
Thermo Scientific Chemicals
Glycerol, 99+%, Extra Pure
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1589200101 LGlass Bottle
Catalog number ACO158920010
Price (SGD)
125.00
Bottle
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
125.00
Bottle
Specifications
Chemical Name or MaterialGlycerol
Name NoteExtra Pure
CAS56-81-5
Decomposition Information>290°C
Melting Point18.0°C