1x1 image pixel for data collection
4-(4-Nitrobenzyl)pyridine, 98%
4-(4-Nitrobenzyl)pyridine, 98%
4-(4-Nitrobenzyl)pyridine, 98%
4-(4-Nitrobenzyl)pyridine, 98%
Thermo Scientific Chemicals
4-(4-Nitrobenzyl)pyridine, 98%
CAS: 1083-48-3 | C12H10N2O2 | 214.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO12855050050 gGlass bottle
Catalog number ACO128550500
Price (SGD)
413.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
413.00
EA
Add to cart
Specifications
Chemical Name or Material4-(4-Nitrobenzyl)pyridine
CAS1083-48-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
RUO – Research Use Only
Share catalog number, name or link