DL-Malic acid, 99+%
DL-Malic acid, 99+%
DL-Malic acid, 99+%
Thermo Scientific Chemicals
DL-Malic acid, 99+%
CAS: 6915-15-7 | C4H6O5 | 134.087 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO125252500250 gPlastic bottle
Catalog number ACO125252500
Price (SGD)
58.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (SGD)
58.00
EA
Add to cart
Specifications
Chemical Name or MaterialDL-Malic acid
CAS6915-15-7
Melting Point128.0°C to 132.0°C
Health Hazard 1Danger
Health Hazard 2GHS P Statement Causes serious eye irritation.