Sodium hydroxide, 1N standard solution
Sodium hydroxide, 1N standard solution
Sodium hydroxide, 1N standard solution
Sodium hydroxide, 1N standard solution
Thermo Scientific Chemicals
Sodium hydroxide, 1N standard solution
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1242600101 LPlastic Bottle
Catalog number ACO124260010
Price (SGD)
69.00
EA
Add to cart
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (SGD)
69.00
EA
Add to cart
Chemical Identifiers
CAS1310-73-2, 7732-18-5
Specifications
Chemical Name or MaterialSodium hydroxide
ColorColorless
CAS1310-73-2
Health Hazard 1Danger
CAS7732-18-5
View more
Volumetric standard solution
RUO – Research Use Only