Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr.
Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr.
Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr.
Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr.
Thermo Scientific Chemicals
Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr.
Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr., Quantity: 500mL, Packaging: Glass bottle, Colorless to Yellow, Molecular Weight: 259.47, Beilstein: 04,II,634, CAS: 2052-49-5, 108-88-3, 67-56-1, Density: 0.8500g/mL, EINECS Number: 218-147-6 | CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO123531000100 mLGlass bottle
Catalog number ACO123531000
Price (SGD)
79.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
79.00
EA
Add to cart
Chemical Identifiers
CAS2052-49-5, 108-88-3, 67-56-1
Specifications
Chemical Name or MaterialTetrabutylammonium hydroxide
CAS2052-49-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes damage to organs.
Suspected of damaging the unborn child.
May be fatal if swallow
CAS108-88-3
View more
Phase transfer catalyst
RUO – Research Use Only