Fluorescein Isothiocyanate Isomer I, 90%, Pure
Fluorescein Isothiocyanate Isomer I, 90%, Pure
Fluorescein Isothiocyanate Isomer I, 90%, Pure
Fluorescein Isothiocyanate Isomer I, 90%, Pure
Thermo Scientific Chemicals
Fluorescein Isothiocyanate Isomer I, 90%, Pure
CAS: 3326-32-7 | C21H11NO5S | 389.381 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACO1192500101 gGlass bottle
Catalog number ACO119250010
Price (SGD)
322.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (SGD)
322.00
EA
Add to cart
Specifications
Chemical Name or MaterialFluorescein isothiocyanate isomer I
CAS3326-32-7
Health Hazard 2GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range90%