1x1 image pixel for data collection
Thermo-Seal Heat Sealing Foil
Thermo-Seal Heat Sealing Foil
Actual product may vary
Thermo-Seal Heat Sealing Foil
Suitable for long-term, low-temperature storage, transportation, and high-temperature applications such as PCR
Have Questions?
Catalog NumberMaterial
AB-0559Aluminum
Catalog number AB-0559
Price (SGD)
171.00
CS
Add to cart
Material:
Aluminum
Price (SGD)
171.00
CS
Add to cart
  • Can be used in waterbath thermal cyclers
  • Foil/polypropylene laminate
  • Provides very strong seal that can be peeled only with the Foil Stripper
  • Can be pierced with a single pipet tip or pierce plate
  • Excellent chemical resistance
  • Seal Integrity Range: -80°C to +120°C
Specifications
ColorSilver
Solvent Or Alcohol ResistantLife Science Reagents Resistant, DMSO Resistant
DescriptionThermo-Seal; foil laminate, 85x135mm
MaterialAluminum
SterilityNon-sterile
AutoclavableNot Autoclavable
No. of Wells96
No. per Case100
No. per Pack100
Dimensions (L x W)85 x 135 mm
For Use With (Application)For ALPS 25 Automated Laboratory Plate Sealer
FormatSheets
For Use With (Equipment)ALPS 25 and ALPS 50V; AB-1443A
Product TypeMicroplate Sealing Components
Product LineAbgene
AdhesivePolypropylene
Certified RNase/DNase-freeNo
PierceableYes
Temperature Range (Metric)-80°C to +120°C
Well ShapeAll well shapes
Plate CompatibilityPolypropylene
Incubation ConditionsGeneral
PeelableVery Strong Seal that requires stripper tool for peeling
Unit SizeCS
Contents & Storage
General; Long term storage including storage at low temperatures
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link