Check Order Status
High-Quality Dehydrants
Actual product may vary
High-Quality Dehydrants
Reduce spills with easy-to-pour Epredia™ High-Quality Dehydrants.
Have Questions?
Catalog Number
Quantity
Product Type
Concentration
9511
1 gal., 4/case
Isopropyl Alcohol
99%
Catalog number
9511
Price (SGD)
784.00
EA
Add to cart
Quantity:
1 gal., 4/case
Product Type:
Isopropyl Alcohol
Concentration:
99%
Price (SGD)
784.00
EA
Add to cart
Reduce spills with easy-to-pour Epredia™ High-Quality Dehydrants.
Formulated to rigid specifications, providing laboratories with high-quality products
Packaged in high-density polyethylene, which is safer and lighter than glass
Specifications
Boiling Point
81° to 83°C
Color
Colorless
Compatibility
Incompatible Materials: Strong oxidizing agents, Acids, Halogens, Acid anhydrides
Concentration
99%
Melting Point
-89.5°C
Product Type
Isopropyl Alcohol
Quantity
1 gal., 4/case
Shelf Life
Hygroscopic
Form
Liquid
Unit Size
EA
Contents & Storage
Keep away from heat, flames and sparks. Exposure to moist air or water.