ClipTip™ Filtered Pipette Tips
ClipTip™ Filtered Pipette Tips
Actual product may vary
ClipTip™ Filtered Pipette Tips
ClipTip™ Filtered Pipette Tips
ClipTip™ Filtered Pipette Tips
Ensure a secure seal for every sample. A unique, innovative interlocking technology ensures a complete seal on every channel with minimal tip attachment and ejection force.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Format
944208131250 μLRack
9442004312.5 μLRack
9442021820 μLReload Deck
9442025850 μLReload Deck
94420318200 μLReload Deck
FIS14-488-008200 μLReload Deck
94420518300 μLReload Deck
944207181000 μLReload Deck
944208181250 μLReload Deck
9442021320 μLRack
FIS14-488-00120 μLRack
FKS1327626920 μLRack
9442025350 μLRack
94420313200 μLRack
FIS14-488-003200 μLRack
FKS13286269200 μLRack
94420513300 μLRack
FIS14-488-004300 μLRack
FKS13296269300 μLRack
944207131000 μLRack
9442006312.5 μLRack
Catalog number 94420813
HSN 84799090
Price (SGD)
146.00
Special offer
Ends: 31-Dec-2025
208.00
Save 62.00 (30%)
PK
Add to cart
Volume (Metric):
1250 μL
Format:
Rack
Price (SGD)
146.00
Special offer
Ends: 31-Dec-2025
208.00
Save 62.00 (30%)
PK
Add to cart
Ensure a secure seal for every sample. Thermo Scientific™ ClipTip™ Pipette Tips provide a secure seal for every sample with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force. Achieve newfound confidence knowing that once attached, your tips are locked firmly in place, and will not loosen or fall off regardless of application pressure. The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip and E1-ClipTip pipettes until — and only until — it is released.
Key Features:
  • ClipTip pipette tips feature 'clips' that lock tips in place and form a complete seal
  • Each time ClipTip pipette tips are on, they are sealed and will not loosen regardless of application pressure
  • ClipTip pipette tips are clipped securely on the F1-ClipTip and E1-ClipTip pipettes with a light touch eliminating the frustration of banging tips on your pipette
  • Filtered ClipTip pipette tips are ideal for sensitive assays, such as PCR, and with critical samples to prevent cross-contaminations
Low retention
  • Low retention technology minimizes liquid retention, and enable maximium sample recovery. Available in ClipTip 20-1250 96-format tips.
Proven quality
  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Environmentally responsible
  • ClipTip reload system uses recycled components and 30% less plastic
Versatile Tip Range
  • 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250μL
  • 384-format volumes: 12.5, 30 and 125μL
  • Extended length tips available for volumes 12.5 and 300 μl allowing for better reach with specific labware consumables
Immediate traceability
  • Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert
Note:
Availability may vary by country.
Specifications
DescriptionClipTip 1250, Filter, Sterile, Rack
Volume (Metric)1250 μL
FilteredFiltered
SterilitySterile
AutoclavableNot Autoclavable
Length (English)4.06 in.
Length (Metric)10.3cm
FormatRack
Tip StyleLow Retention
Packaging8 x 96 tips/rack
Quantity768 tips
Color CodeTurquoise
For Use With (Equipment)Thermo Scientific E1-ClipTip, Thermo Scientific F1-ClipTip
Product LineClipTip
Fit for PurposeSterile & PCR Ready
Unit SizePK