Search Thermo Fisher Scientific
The Thermo Scientific™ Finntip™ Flex™ Pipette Tips reduce the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI). Ensure a better seal with the pipetter module, for higher accuracy and precision.
Catalog Number | Volume (Metric) | Color Code | Fit for Purpose |
---|---|---|---|
94060320 | 200 μL | Yellow | Certified Quality |
94060120 | 10 μL | Pink | Certified Quality |
94060100 | 10 μL | Pink | Certified Quality |
FIS14-387-208 | 10 μL | Pink | Certified Quality |
94060310 | 200 μL | Yellow | Certified Quality |
FIS14-387-200 | 200 μL | Yellow | Certified Quality |
94060103 | 10 μL | Pink | Sterile & PCR Ready |
FIS14-387-209 | 10 μL | Pink | Sterile & PCR Ready |
94060313 | 200 μL | Yellow | Sterile & PCR Ready |
FIS14-387-201 | 200 μL | Yellow | Sterile & PCR Ready |
94060316 | 200 μL | Yellow | Non-sterile |
94060117 | 10 μL | Pink | Certified Quality |
94060317 | 200 μL | Yellow | Certified Quality |
FIS14-387-204 | 200 μL | Yellow | Certified Quality |
94060510 | 300 μL | Orange | Certified Quality |
94060513 | 300 μL | Orange | Sterile & PCR Ready |
94060517 | 300 μL | Orange | Certified Quality |
94060710 | 1000 μL | Blue | Certified Quality |
FIS14-387-84 | 1000 μL | Blue | Certified Quality |
94060713 | 1000 μL | Blue | Sterile & PCR Ready |
FIS14-387-85 | 1000 μL | Blue | Sterile & PCR Ready |
94060716 | 1000 μL | Blue | |
94060717 | 1000 μL | Blue | Certified Quality |
FIS14-387-87 | 1000 μL | Blue | Certified Quality |
94060720 | 1000 μL | Blue | Certified Quality |
94060810 | 1200 μL | Turquoise | Certified Quality |
FIS14-387-82 | 1200 μL | Turquoise | Certified Quality |
94060813 | 1200 μL | Turquoise | Sterile & PCR Ready |
94060817 | 1200 μL | Turquoise | Certified Quality |
94060820 | 1200 μL | Turquoise | Certified Quality |
Finntip™ Flex™ Pipette Tips are designed to fit Finnpipette™ F1, F2 and Novus pipettes. They enable you to pipette accurately and precisely, with low liquid retention. The compatibility between the tip and the pipette is indicated with color codes. These Flex™ tips have an ergonomic design that lower the risk of RSI and use less plastic.
Key Features:
Softer Finntip Flex Tip
Proven quality
Versatile Tip Range
Immediate traceability
Note:
Availability may vary by country.